(S)-2-Amino-3,3-dimethylbutanoic acid hydrochloride 95%

Supplier: Ambeed
Warning

A250366-100G A250366-1G A250366-25G A250366-5G
76801-536EA 174 USD
76801-536 76801-538 76801-540 76801-542
(S)-2-Amino-3,3-dimethylbutanoic acid hydrochloride 95%
(S)-2-Amino-3,3-dimethylbutanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MDL Number: MFCD07368368
CAS Number: 139163-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR