Diethyl-sec-butylmalonate 95%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-(sec-butyl)malonate

A250078-1G A250078-25G A250078-5G
76801-530EA 5.31 USD
76801-530 76801-532 76801-534
Diethyl-sec-butylmalonate 95%
Diethyl-sec-butylmalonate

Precaución: for Research Use Only

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
MDL Number: MFCD00015194
CAS Number: 83-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR