8-Quinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Quinoline-8-carboxylic acid

A248196-10G A248196-1G A248196-250MG A248196-25G A248196-5G A248196-100G
76801-508EA 150.65 USD
76801-508 76801-510 76801-512 76801-514 76801-516 77626-308
8-Quinolinecarboxylic acid 98%
8-Quinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
MDL Number: MFCD00047619
CAS Number: 86-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR