Dithieno[3,2-b:2',3'-d]thiophene 98%
Supplier: Ambeed
|
Warning
|
A247823-100MG
A247823-1G
A247823-250MG
A247823-5G
76801-496EA
23.34
USD
76801-496
76801-498
76801-500
76801-502
Dithieno[3,2-b:2',3'-d]thiophene 98%
Dithieno[3,2-b:2',3'-d]thiophene
Precaución: for Research Use Only
Formula:
C₈H₄S₃ MW: 196.30 g/mol |
MDL Number:
MFCD06656577 CAS Number: 3593-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...