1-(2,4-Dichlorophenyl)cyclopropanecarboxylic acid 98%

Supplier: Ambeed
Warning

A247557-1G A247557-250MG A247557-5G
76801-490EA 80.63 USD
76801-490 76801-492 76801-494
1-(2,4-Dichlorophenyl)cyclopropanecarboxylic acid 98%
1-(2,4-Dichlorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₂
MW: 231.0800 g/mol
MDL Number: MFCD00044817
CAS Number: 84604-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR