1-Bromo-2-(bromomethyl)naphthalene 98%

Supplier: Ambeed
Danger

A247341-250MG A247341-25G A247341-1G
76801-486EA 25 USD
76801-486 76801-488 77879-451
1-Bromo-2-(bromomethyl)naphthalene 98%
1-Bromo-2-(bromomethyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₁H₈Br₂
MDL Number: MFCD00046369
CAS Number: 37763-43-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR