DL-Valinol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-methylbutan-1-ol, DL-2-Amino-3-methyl-1-butanol, 2-Amino-3-methyl-1-butanol

A247012-100G A247012-10G A247012-1G A247012-250MG A247012-5G
76801-468EA 363 USD
76801-468 76801-470 76801-472 76801-474 76801-476
DL-Valinol 97%
DL-Valinol

Precaución: for Research Use Only

Formula: C₅H₁₃NO
MW: 103.17 g/mol
MDL Number: MFCD00004730
CAS Number: 16369-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR