(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
D-Phenylalanine tert-butyl ester hydrochloride, H-D-Phe-OtBu·HCl, H-D-Phe-OtBu.HCl
A246896-10G
A246896-1G
A246896-5G
A246896-100G
A246896-25G
76801-462EA
22
USD
76801-462
76801-464
76801-466
77600-992
77668-010
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride 97%
(R)-tert-Butyl-2-amino-3-phenylpropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₉NO₂·HCl |
MDL Number:
MFCD00153455 CAS Number: 3403-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...