8-(Trifluoromethyl)-4-quinolinol 97%

Supplier: Ambeed
Warning

A246763-10G A246763-1G A246763-5G
76801-456EA 91 USD
76801-456 76801-458 77879-446
8-(Trifluoromethyl)-4-quinolinol 97%
8-(Trifluoromethyl)-4-quinolinol

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
MDL Number: MFCD00134577
CAS Number: 23779-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR