Quinaldic acid ≥99%

Supplier: Ambeed
Warning

A246501-100G A246501-10G A246501-25G A246501-5G A246501-500G
76801-444EA 94 USD
76801-444 76801-446 76801-448 76801-450 77600-984
Quinaldic acid ≥99%
Quinaldic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MDL Number: MFCD00006752
CAS Number: 93-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR