Fmoc-Phe-OPfp 96%

Supplier: Ambeed
Warning

A246006-1G A246006-25G A246006-5G A246006-100G
76801-430EA 17 USD
76801-430 76801-432 76801-434 77879-442
Fmoc-Phe-OPfp 96%
Fmoc-Phe-OPfp

Precaución: for Research Use Only

Formula: C₃₀H₂₀F₅NO₄
MDL Number: MFCD00065669
CAS Number: 86060-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR