1-Benzyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl 3-ethyl 4-oxopyrrolidine-1,3-dicarboxylate

A245051-100G A245051-10G A245051-1G A245051-250MG A245051-25G A245051-5G
76801-414EA 931.48 USD
76801-414 76801-416 76801-418 76801-420 76801-422 76801-424
1-Benzyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate 95%
1-Benzyl-3-ethyl-4-oxopyrrolidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO₅
MW: 291.31 g/mol
MDL Number: MFCD09878816
CAS Number: 51814-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR