2-(3-Phenoxyphenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A244887-100MG A244887-1G A244887-250MG A244887-5G
76801-400EA 26.53 USD
76801-400 76801-402 76801-404 76801-406
2-(3-Phenoxyphenyl)propanoic acid 98%
2-(3-Phenoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00072027
CAS Number: 29679-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR