H-Tyr(tBu)-OMe·HCl 95%

Supplier: Ambeed
Warning

Synonyms: Methyl (S)-2-amino-3-(4-(tert-butoxy)phenyl)propanoate hydrochloride, H-Tyr(tBu)-OMe·HCl

A243912-10G A243912-1G A243912-250MG A243912-25G
76801-386EA 56.23 USD
76801-386 76801-388 76801-390 76801-392
H-Tyr(tBu)-OMe·HCl 95%
H-Tyr(tBu)-OMe·HCl

Precaución: for Research Use Only

Formula: C₁₄H₂₂ClNO₃
MW: 287.784 g/mol
MDL Number: MFCD00153466
CAS Number: 51482-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR