H-Asp(Obzl)-OtBu.HCl 97%

Supplier: Ambeed
Warning

A242703-100G A242703-10G A242703-25G A242703-5G
76801-370EA 292.81 USD
76801-370 76801-372 76801-374 76801-376
H-Asp(Obzl)-OtBu.HCl 97%
H-Asp(Obzl)-OtBu.HCl

Precaución: for Research Use Only

Formula: C₁₅H₂₂ClNO₄
MW: 315.79 g/mol
MDL Number: MFCD00190761
CAS Number: 52615-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR