2-(4-Chlorophenyl)-3-methylbutyric acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chlorophenyl)-3-methylbutanoic acid

A242628-100G A242628-25G A242628-5G
76801-366EA 55.17 USD
76801-366 76801-368 77667-980
2-(4-Chlorophenyl)-3-methylbutyric acid 98%
2-(4-Chlorophenyl)-3-methylbutyric acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.67 g/mol
MDL Number: MFCD00037763
CAS Number: 2012-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR