EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)] 97%

Supplier: Ambeed
Warning

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) O,O'-ethane-1,2-diyl disuccinate, EGS, Ethylene glycol disuccinate di(N-succinimidyl) ester, Ethylene glycol bis(succinimidyl succinate), Di(N-succinimidyl) ethylene glycol disuccinate

A240846-100MG A240846-1G A240846-250MG A240846-5G
76801-322EA 33 USD
76801-322 76801-324 76801-326 76801-328
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)] 97%
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O₁₂
MW: 456.36 g/mol
MDL Number: MFCD00049057
CAS Number: 70539-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR