Guanosine-5'-diphosphate sodium salt 97%

Supplier: Ambeed
Warning

Synonyms: Sodium ((2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate, Guanosine 5'-diphosphate sodium

A240704-1G A240704-5G
76801-320EA 71.09 USD
76801-320 77568-334
Guanosine-5'-diphosphate sodium salt 97%
Guanosine 5'-diphosphate sodium salt

Precaución: For research use only.

Formula: C₁₀H₁₄N₅NaO₁₁P₂
MW: 509.1460 g/mol
MDL Number: MFCD00070121
CAS Number: 43139-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR