2-Methylquinoline-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A239245-10G A239245-1G A239245-25G A239245-5G
76801-298EA 126.25 USD
76801-298 76801-300 76801-302 76801-304
2-Methylquinoline-4-carboxylic acid 98%
2-Methylquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD00093919
CAS Number: 634-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR