2-(4-Isobutylphenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: (±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen

A238702-500G A238702-5G A238702-25G A238702-1000G A238702-100G A238702-1MG
76801-290EA 58.35 USD
76801-290 76801-292 77090-162 77568-316 77568-318 77667-952
2-(4-Isobutylphenyl)propanoic acid 98%
2-(4-Isobutylphenyl)propanoic acid

Precaución: For research use only.

Formula: C₁₃H₁₈O₂
MW: 206.3 g/mol
MDL Number: MFCD00010393
CAS Number: 15687-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR