Ethyl 5-amino-1-(tert-butyl)-1H-pyrazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A238371-100MG A238371-1G A238371-250MG
76801-284EA 9.55 USD
76801-284 76801-286 76801-288
Ethyl 5-amino-1-(tert-butyl)-1H-pyrazole-4-carboxylate 97%
Ethyl 5-amino-1-(tert-butyl)-1H-pyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₇N₃O₂
MW: 211.26 g/mol
MDL Number: MFCD06637286
CAS Number: 112779-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR