Dimethyl-5-nitroisophthalate 97%

Supplier: Ambeed
Warning

Synonyms: Dimethyl 5-nitroisophthalate, 5-Nitroisophthalic acid dimethyl ester

A238281-1000G A238281-100G A238281-25G A238281-500G
76801-276EA 78.51 USD
76801-276 76801-278 76801-280 76801-282
Dimethyl-5-nitroisophthalate 97%
Dimethyl-5-nitroisophthalate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MW: 239.18 g/mol
MDL Number: MFCD00008429
CAS Number: 13290-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR