DL-α-Tocopherol (Vitamin E) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E
A238186-25G
A238186-500G
A238186-1000G
A238186-100G
76801-272EA
24
USD
76801-272
76801-274
77550-536
77568-302
DL-α-Tocopherol (Vitamin E) 98%
DL-α-Tocopherol (Vitamin E)
Precaución: For research use only.
Formula:
C₂₉H₅₀O₂ MW: 430.71 g/mol |
MDL Number:
MFCD00072051 CAS Number: 10191-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...