Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-Ethyl 3-(pyrrolidin-1-yl)but-2-enoate
A237600-100G
A237600-10G
A237600-1G
A237600-25G
A237600-5G
76801-252EA
292.81
USD
76801-252
76801-254
76801-256
76801-258
76801-260
Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%
Ethyl (E)-3-(1-pyrrolidinyl)crotonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₂ MW: 183.25 g/mol |
MDL Number:
MFCD00014097 CAS Number: 54716-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...