Isobutyl benzoate 98%

Supplier: Ambeed
Warning

A237568-100G A237568-25G A237568-500G
76801-246EA 42 USD
76801-246 76801-248 76801-250
Isobutyl benzoate 98%
Isobutyl benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD00048344
CAS Number: 120-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR