Methyl-2,3-O-isopropylidene-β-D-ribofuranoside 95%

Supplier: Ambeed
Warning

Synonyms: ((3aR,4R,6R,6aR)-6-Methoxy-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)methanol

A237164-100G A237164-10G A237164-1G A237164-250MG A237164-25G A237164-5G
76801-228EA 457.25 USD
76801-228 76801-230 76801-232 76801-234 76801-236 76801-238
Methyl-2,3-O-isopropylidene-β-D-ribofuranoside 95%
Methyl-2,3-O-isopropylidene-beta-D-ribofuranoside

Precaución: for Research Use Only

Formula: C₉H₁₆O₅
MW: 204.222 g/mol
MDL Number: MFCD00270040
CAS Number: 4099-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR