Ethyl 2-(piperidin-4-yl)acetate hydrochloride 97%

Supplier: Ambeed
Warning

A236026-10G A236026-1G A236026-5G
76801-210EA 195.21 USD
76801-210 76801-212 76801-214
Ethyl 2-(piperidin-4-yl)acetate hydrochloride 97%
Ethyl 2-(piperidin-4-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₂
MW: 207.70 g/mol
MDL Number: MFCD08703262
CAS Number: 169458-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR