Malonamamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Malonamideamidine hydrochloride

A235840-100G A235840-10G A235840-25G A235840-500G A235840-5G
76801-198EA 108 USD
76801-198 76801-200 76801-202 76801-204 76801-206
Malonamamidine hydrochloride 97%
Malonamamidine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇N₃O·HCl
MW: 137.57 g/mol
MDL Number: MFCD00013007
CAS Number: 34570-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR