Ethyl-2-amino-4-fluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4-fluorobenzoate

A235769-10G A235769-1G A235769-250MG
76801-192EA 233.4 USD
76801-192 76801-194 76801-196
Ethyl-2-amino-4-fluorobenzoate 98%
Ethyl-2-amino-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Number: MFCD08275015
CAS Number: 117324-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR