Diamyl disulfide 98%

Supplier: Ambeed
Warning

Synonyms: Di-n-amyldisulphide, Pentyl Disulphide, Dipentyl Disulphide, Amyl Disulphide

A235346-100G A235346-25G A235346-500G
76801-180EA 19.1 USD
76801-180 76801-182 76801-184
Diamyl disulfide 98%
Diamyl disulfide

Precaución: for Research Use Only

Formula: C₁₀H₂₂S₂
MW: 206.41 g/mol
MDL Number: MFCD00039964
CAS Number: 112-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR