Methyl benzimidazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Benzimidazole-5-carboxylic acid methyll ester

A235131-100G A235131-10G A235131-1G A235131-25G A235131-5G
76801-158EA 323.58 USD
76801-158 76801-160 76801-162 76801-164 76801-166
Methyl benzimidazole-5-carboxylate 98%
Methyl benzimidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.1720 g/mol
MDL Number: MFCD03407310
CAS Number: 26663-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR