4-Isobutoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A234252-100G A234252-10G A234252-1G A234252-250MG A234252-25G
76801-144EA 657.76 USD
76801-144 76801-146 76801-148 76801-150 76801-152
4-Isobutoxybenzaldehyde 95%
4-Isobutoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01922006
CAS Number: 18962-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR