Inosine-5-diphosphate disodium salt 97%

Supplier: Ambeed
Warning

Synonyms: Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl hydrogen diphosphate

A234111-10G A234111-1G A234111-5G
76801-138EA 85.94 USD
76801-138 76801-140 76801-142
Inosine-5-diphosphate disodium salt 97%
Inosine-5-diphosphate disodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄Na₂O₁₁P₂
MW: 472.15 g/mol
MDL Number: MFCD00211028
CAS Number: 54735-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR