7-Nitro-1-tetralone 97%
Supplier: Ambeed
|
Warning
|
A233041-100G
A233041-1G
A233041-250MG
A233041-25G
A233041-5G
76801-122EA
341
USD
76801-122
76801-124
76801-126
76801-128
76801-130
7-Nitro-1-tetralone 97%
7-Nitro-1-tetralone
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₃ |
MDL Number:
MFCD00019661 CAS Number: 40353-34-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...