Linoleic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

A232282-5G A232282-100G A232282-25G A232282-500G
76801-110EA 27 USD
76801-110 77550-504 77550-506 77593-506
Linoleic acid ≥99%
Linoleic acid

Precaución: for Research Use Only

Formula: C₁₈H₃₂O₂
MDL Number: MFCD00064241
CAS Number: 60-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR