Dimethyl itaconate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester
A231564-100G
A231564-10G
A231564-500G
A231564-25G
76801-100EA
16
USD
76801-100
76801-102
77550-502
77600-860
Dimethyl itaconate 98%
Dimethyl itaconate
Precaución: for Research Use Only
Formula:
C₇H₁₀O₄ |
MDL Number:
MFCD00008446 CAS Number: 617-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...