Zinc 2-ethylhexanoate Zn ~18%

Supplier: Ambeed
Warning

Synonyms: Zinc(II) 2-ethylhexanoate

A230019-100G A230019-500G A230019-25G
76801-072EA 9 USD
76801-072 76801-074 77568-228
Zinc 2-ethylhexanoate Zn ~18%
Zinc 2-ethylhexanoate

Precaución: For research use only.

Formula: C₁₆H₃₀O₄Zn
MW: 351.792 g/mol
MDL Number: MFCD00053316
CAS Number: 136-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR