Methyl 2-(5-bromo-2-hydroxyphenyl)acetate 98%

Supplier: Ambeed
Warning

A229500-1G A229500-250MG A229500-5G
76801-062EA 25 USD
76801-062 76801-064 76801-066
Methyl 2-(5-bromo-2-hydroxyphenyl)acetate 98%
Methyl 2-(5-bromo-2-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD07778484
CAS Number: 220801-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR