Bis(lauroyloxy)dioctyltin 96%

Supplier: Ambeed
Danger

Synonyms: [Dodecanoyloxy(dioctyl)stannyl] dodecanoate

A229321-100G A229321-25G A229321-500G
76801-056EA 54 USD
76801-056 76801-058 76801-060
Bis(lauroyloxy)dioctyltin 96%
Bis(lauroyloxy)dioctyltin

Precaución: for Research Use Only

Formula: C₄₀H₈₀O₄Sn
MDL Number: MFCD00026557
CAS Number: 3648-18-8
UN: 2788
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR