Ethyl salicylate 98%

Supplier: Ambeed
Warning

A229111-1000G A229111-100G A229111-500G
76801-050EA 39 USD
76801-050 76801-052 76801-054
Ethyl salicylate 98%
Ethyl salicylate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00002215
CAS Number: 118-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR