1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone 97%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-4-(p-tolylamino)anthracene-9,10-dione

A228703-500G A228703-100G A228703-25G
76801-034EA 73.21 USD
76801-034 77568-216 77600-830
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone 97%
1-Hydroxy-4-(p-toluidino)-9,10-anthraquinone

Precaución: For research use only.

Formula: C₂₁H₁₅NO₃
MW: 329.3500 g/mol
MDL Number: MFCD00019148
CAS Number: 81-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR