Ethyl-3-methylvalerate 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl 3-methylpentanoate, Ethyl 3-Methylpentanoate, 3-Methylpentanoic Acid Ethyl Ester, 3-Methylvaleric Acid Ethyl Ester

A228301-25G A228301-5G
76801-024EA 118 USD
76801-024 76801-026
Ethyl-3-methylvalerate 98%
Ethyl-3-methylvalerate

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MDL Number: MFCD00041390
CAS Number: 5870-68-8
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR