4-Methylbenzoylacetonitrile 98%

Supplier: Ambeed
Warning

A226872-100G A226872-1G A226872-25G A226872-5G
76800-994EA 303 USD
76800-994 76800-996 76800-998 76801-000
4-Methylbenzoylacetonitrile 98%
4-Methylbenzoylacetonitrile

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD00067895
CAS Number: 7391-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR