2-Ethoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol 97% (by GC) contains isomer

Supplier: Ambeed
Warning

A226554-100G A226554-25G
76800-986EA 51 USD
76800-986 76800-988
2-Ethoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol 97% (by GC) contains isomer
2-Ethoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MDL Number: MFCD00036667
CAS Number: 68527-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR