Forskolin 98%

Supplier: Ambeed
Warning

A226193-100MG A226193-10MG A226193-1G A226193-1MG A226193-250MG A226193-50MG A226193-5MG A226193-5G
76800-972EA 71 USD
76800-972 76800-974 76800-976 76800-978 76800-980 76800-982 76800-984 77600-812
Forskolin 98%
Forskolin

Precaución: for Research Use Only

Formula: C₂₂H₃₄O₇
MDL Number: MFCD00082317
CAS Number: 66575-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR