3-(1-Cyanoethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A225628-1000G A225628-100G A225628-10G A225628-25G A225628-500G A225628-5G
76800-960EA 405.01 USD
76800-960 76800-962 76800-964 76800-966 76800-968 76800-970
3-(1-Cyanoethyl)benzoic acid 97%
3-(1-Cyanoethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00002520
CAS Number: 5537-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR