Kaempferitrin 95%

Supplier: Ambeed
Warning

A224689-100MG A224689-10MG A224689-50MG
76800-936EA 374 USD
76800-936 76800-938 76800-940 76800-942 77879-362
Kaempferitrin 95%
Kaempferitrin

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₁₄
MDL Number: MFCD01662671
CAS Number: 482-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR