6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-(4-Aminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one, Levosimendan dazinone intermediate
A223245-100MG
A223245-1G
A223245-250MG
A223245-5G
76800-918EA
5.31
USD
76800-918
76800-920
76800-922
76800-924
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 97%
6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone
Precaución: for Research Use Only
Formula:
C₁₁H₁₃N₃O MW: 203.25 g/mol |
MDL Number:
MFCD01692712 CAS Number: 36725-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...