Methyl-2-hydroxy-1-naphthoate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-1-naphthoic acid methyl ester, 2-Hydroxy-α-naphthoic acid methyl ester, Methyl 2-hydroxy-1-naphthoate

A223222-10G A223222-1G A223222-25G A223222-5G
76800-910EA 222.79 USD
76800-910 76800-912 76800-914 76800-916
Methyl-2-hydroxy-1-naphthoate 95%
Methyl-2-hydroxy-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Number: MFCD00454146
CAS Number: 947-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR