3-(Ethoxycarbonyl)phenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
A221980-10G
A221980-1G
A221980-25G
A221980-5G
76800-898EA
60.48
USD
76800-898
76800-900
76800-902
76800-904
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester 98%
3-(Ethoxycarbonyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₁BO₄ MW: 276.136 g/mol |
MDL Number:
MFCD03093896 CAS Number: 269410-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...